Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Side Access Freezer Rack, Holds (36) 50-Cell Hinged Top Boxes, Rack Dims 24 1/16L x 14 1/8H x 5 7/8W" (611 x 359 x 150mm), Compartment Dims 3 9/16L x 2 5/16H x 5 9/16W" (91 x 58 x 142mm) 6x6
Catalog No. NC2208104
Encompass_Preferred
Rack Designed to hold (36) 50-Cell Hinged Top Plastic Boxes (2 1/5"H x5 9/16W x3 3/8L" (56 x 141 x 86mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208104 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFH5066
Add to Cart
Add to Cart
Rack Designed to hold (36) 50-Cell Hinged Top Plastic Boxes (2 1/5"H x5 9/16W x3 3/8L" (56 x 141 x 86mm), made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.