Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Upright Side Access Rack Holds (84) Plates w/Lid(96)no Lids Rack Dims 22 1/2L x 9 3/16H x 5 1/2W"(571 x 233 x 140m)Compartment Dims 3 9/16L x 4 1/2H x 5 13/16W"(91 x 115 x 131mm)Plate H 9/16"(14.3mm)
Catalog No. NC2208100
Encompass_Preferred
Rack Designed to hold 96 Well and 384 Well Plates (84) Plates with lids/(96) Plates no lids, Locking Rods Included, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.
This item is not returnable. View return policy
Catalog No. NC2208100 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. UFMP609L
Rack Designed to hold 96 Well and 384 Well Plates (84) Plates with lids/(96) Plates no lids, Locking Rods Included, made of corrosion resistant stainless steel, with Lifetime Warranty, Double Welds and Smooth Edges, many other sizes of Upright and Chest Freezer Racks as well as Canisters and Frames for LN2 Tanks available. Custom Rack Design Services available. Security lock devices available for each rack.