Learn More
Alkali Scientific Alkali Scientific™ Upright Sliding Tray Freezer Rack for 3" Boxes, 4 x 3 Configuration, 12 Box Capacity
SDP
Catalog No. 503027995
Encompass_Preferred
For Standard 3″ Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions L x H x W: 22 3/8 x 9 1/2 x 5 1/2 inch (568 x 241 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 1/16 x 5 1/4 (137.4 x 76.7 x 134mm)
Catalog No. 50-302-7995 Supplier Alkali Scientific Supplier No. UFS433
For Standard 3″ Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions L x H x W: 22 3/8 x 9 1/2 x 5 1/2 inch (568 x 241 x 140mm)Compartment Size L x H x W: 5 3/8 x 3 1/16 x 5 1/4 (137.4 x 76.7 x 134mm)