Learn More
Alkali Scientific Alkali Scientific™ Upright Sliding Tray Freezer Rack for 2" Boxes, 3 x 5 Configuration, 15 Box Capacity
SDP
Catalog No. 503028012
Encompass_Preferred
For Standard 2″ Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions (LxHxW): 16 7/8 x 11 x 5 1/2 (428 x 168 x 140mm)Compartment Dimensions: 5 3/8 x 2 1/16 x 5 1/4 inch (137.4 x 53 x 134.1mm)
Catalog No. 50-302-8012 Supplier Alkali Scientific Supplier No. UFS352
For Standard 2″ Boxes Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.   Features: Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Dimensions (LxHxW): 16 7/8 x 11 x 5 1/2 (428 x 168 x 140mm)Compartment Dimensions: 5 3/8 x 2 1/16 x 5 1/4 inch (137.4 x 53 x 134.1mm)