Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Rack for 3" Boxes, 3 x 5 Configuration, 15 Box Capacity
SDP
Catalog No. 503028002
Encompass_Preferred
For Standard 2″ Boxes3×5 Configuration Standard upright freezer racks for storage of 3 inch boxes, features a 3 boxes deep by 5 boxes high configuration for a total storage of 15 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxWxH): 16 7/8 x 5 1/2 x 15 9/16 inches (429 x 140 x 395mm)Compartment Dimensions: 5 3/8 x 5 1/2 x 3 1/16 inches (137 x 140 x 78mm)
Catalog No. 50-302-8002 Supplier Alkali Scientific Supplier No. UFR353
For Standard 2″ Boxes3×5 Configuration Standard upright freezer racks for storage of 3 inch boxes, features a 3 boxes deep by 5 boxes high configuration for a total storage of 15 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxWxH): 16 7/8 x 5 1/2 x 15 9/16 inches (429 x 140 x 395mm)Compartment Dimensions: 5 3/8 x 5 1/2 x 3 1/16 inches (137 x 140 x 78mm)