Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Rack for 2" Boxes, 3 x 7 Configuration, 21 Box Capacity
SDP
Catalog No. 503028037
Encompass_Preferred
For Standard 2″ Boxes3×7 Configuration Standard upright freezer racks for storage of 2 inch boxes, features a 3 boxes deep by 7 boxes high configuration for a total storage of 21 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxHxW): 16 7/8 x 15 3/8 x 5 1/2 inches (429 x 391 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)
Catalog No. 50-302-8037 Supplier Alkali Scientific Supplier No. UFR372
For Standard 2″ Boxes3×7 Configuration Standard upright freezer racks for storage of 2 inch boxes, features a 3 boxes deep by 7 boxes high configuration for a total storage of 21 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxHxW): 16 7/8 x 15 3/8 x 5 1/2 inches (429 x 391 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)