Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Racks for 2” Boxes- Rack Only- 2 Boxes Deep x 7 Boxes High
SDP
Catalog No. 503027572
Encompass_Preferred
For Standard 2″ Boxes2×7 Configuration Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 11 1/2 x 15 3/8 x 5 1/2 inches (292 x 391 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)
Catalog No. 50-302-7572 Supplier Alkali Scientific Supplier No. UFR272
For Standard 2″ Boxes2×7 Configuration Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 11 1/2 x 15 3/8 x 5 1/2 inches (292 x 391 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)