Learn More
Alkali Scientific Upright Stainless-Steel Freeze
SDP
Catalog No. 503027573
Encompass_Preferred
The Alkali ScientificUpright Stainless Steel Freezer Rack for Mixed Storage is a versatile and robust solution designed to accommodate a variety of standard box sizes including 2in 3in and 3 75in heights This innovative rack system is ideal for laboratories seeking a flexible storage option that can adapt to different sample sizes and types all while ensuring optimal organization and accessibility within upright freezers Product Features Versatile Storage CompatibilitySpecifically engineered to hold a mix of box heights this freezer rack allows for the organized storage of 2in 3in and 3 75in standard boxes Its adaptable design facilitates the efficient management of diverse sample collections making it perfect for laboratories with a wide range of storage needs Premium Stainless-Steel ConstructionCrafted from high-quality stainless steel this upright freezer rack offers unmatched durability and resistance to the harsh conditions of freezer environments The stainless-steel material
Catalog No. 50-302-7573 Supplier Alkali Scientific Supplier No. UFR406
The Alkali ScientificUpright Stainless Steel Freezer Rack for Mixed Storage is a versatile and robust solution designed to accommodate a variety of standard box sizes including 2in 3in and 3 75in heights This innovative rack system is ideal for laboratories seeking a flexible storage option that can adapt to different sample sizes and types all while ensuring optimal organization and accessibility within upright freezers Product Features Versatile Storage CompatibilitySpecifically engineered to hold a mix of box heights this freezer rack allows for the organized storage of 2in 3in and 3 75in standard boxes Its adaptable design facilitates the efficient management of diverse sample collections making it perfect for laboratories with a wide range of storage needs Premium Stainless-Steel ConstructionCrafted from high-quality stainless steel this upright freezer rack offers unmatched durability and resistance to the harsh conditions of freezer environments The stainless-steel material