Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Freezer Rack for Mixed Storage of 2″,3″ and 3.75″ Standard Boxes, 27 3/16 x 12 3/8 x 51 1/2"
SDP
Catalog No. 503027574
Encompass_Preferred
For Mixed Storage of  2″,3″ and 3.75″ Standard Boxes The Alkali Scientific™ Upright Stainless Steel Freezer Rack for Mixed Storage is a versatile and robust solution designed to accommodate a variety of standard box sizes, including 2", 3", and 3.75" heights. This innovative rack system is ideal for laboratories seeking a flexible storage option that can adapt to different sample sizes and types, all while ensuring optimal organization and accessibility within upright freezers. Dimensions L x H x W: 27 3/16 x 12 3/8 x 51 1/2 inch( 691 x 315 x 140mm)Compartment Dimensions: 5 5/16 x 12 3/16 x 5 7/16 inch ( 134.7 x 310 x 138mm)
Catalog No. 50-302-7574 Supplier Alkali Scientific Supplier No. UFR512L
For Mixed Storage of  2″,3″ and 3.75″ Standard Boxes The Alkali Scientific™ Upright Stainless Steel Freezer Rack for Mixed Storage is a versatile and robust solution designed to accommodate a variety of standard box sizes, including 2", 3", and 3.75" heights. This innovative rack system is ideal for laboratories seeking a flexible storage option that can adapt to different sample sizes and types, all while ensuring optimal organization and accessibility within upright freezers. Dimensions L x H x W: 27 3/16 x 12 3/8 x 51 1/2 inch( 691 x 315 x 140mm)Compartment Dimensions: 5 5/16 x 12 3/16 x 5 7/16 inch ( 134.7 x 310 x 138mm)