Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Racks for 3” Boxes- Rack Only- 2 Boxes Deep x 5 Boxes High
SDP
Catalog No. 503027907
Encompass_Preferred
For Standard 3″ Boxes2×5 Configuration Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 11 1/2 x 5 1/2 x 15 9/16 inches (292 x 140 x 395mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (136 x 138 x 78mm)
Catalog No. 50-302-7907 Supplier Alkali Scientific Supplier No. UFR253
For Standard 3″ Boxes2×5 Configuration Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 11 1/2 x 5 1/2 x 15 9/16 inches (292 x 140 x 395mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 3 1/16 inches (136 x 138 x 78mm)