Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Rack for 3" Boxes, 5 x 2 Configuration, 10 Box Capacity
SDP
Catalog No. 503027943
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack 5 Long x 2 Tall= Holds 10 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 27 5/16 x 5 1/2 x 6 5/8 inches (693 x 140 x 168mm)Compartment Dimensions: 5 7/16 x 5 1/4 x 3 1/4 inches (134 x 138 x 82.5mm)
Catalog No. 50-302-7943 Supplier Alkali Scientific Supplier No. UFR523
Constructed of corrosion resistant stainless steel Racks fit standard 3" cardboard, plastic, or metal boxes Security lock devices available for each rack 5 Long x 2 Tall= Holds 10 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxWxH): 27 5/16 x 5 1/2 x 6 5/8 inches (693 x 140 x 168mm)Compartment Dimensions: 5 7/16 x 5 1/4 x 3 1/4 inches (134 x 138 x 82.5mm)