Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Freezer Rack (UFH50-53) for 50-Cell Hinged Top Plastic Boxes, 5 x 3 Configuration, 15 Box Capacity
SDP
Catalog No. 503028080
Encompass_Preferred
For 50-Cell Hinged Top Plastic Boxes The Alkali Scientific™ Upright Stainless Steel Freezer Rack (UFH50-53) for 50-Cell Hinged Top Plastic Boxes is a premium storage solution designed to provide unparalleled organization and accessibility for samples stored in 50-cell hinged top plastic boxes within upright freezer environments. Crafted from high-quality stainless steel, this freezer rack combines durability with efficient space management, making it an ideal choice for laboratories seeking to optimize their cold storage practices. Dimensions (DxWxH): 18 13/16 x 5 7/8 x 7 1/4 inches (478 x 150 x 184mm)Compartment Dimensions:3 9/16 x 2 5/16 x 5 9/16 inch (91 x 58 x 142mm)
Catalog No. 50-302-8080 Supplier Alkali Scientific Supplier No. UFH5053
For 50-Cell Hinged Top Plastic Boxes The Alkali Scientific™ Upright Stainless Steel Freezer Rack (UFH50-53) for 50-Cell Hinged Top Plastic Boxes is a premium storage solution designed to provide unparalleled organization and accessibility for samples stored in 50-cell hinged top plastic boxes within upright freezer environments. Crafted from high-quality stainless steel, this freezer rack combines durability with efficient space management, making it an ideal choice for laboratories seeking to optimize their cold storage practices. Dimensions (DxWxH): 18 13/16 x 5 7/8 x 7 1/4 inches (478 x 150 x 184mm)Compartment Dimensions:3 9/16 x 2 5/16 x 5 9/16 inch (91 x 58 x 142mm)