Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Freezer Rack for 2" Polycarbonate Boxes, 4x6 Configuration, 24 Box Capacity
SDP
Catalog No. 503027847
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 6 Tall= Holds 24 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (LxHxW): 21.75 x 13.08 x 5.5 inches (554 x 333 x 140 mm)Compartment Internal Dimensions (LxHxW): 54.98 x 133.5 x 137.7 mm
Catalog No. 50-302-7847 Supplier Alkali Scientific Supplier No. UFR462P
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack 4 Long x 6 Tall= Holds 24 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (LxHxW): 21.75 x 13.08 x 5.5 inches (554 x 333 x 140 mm)Compartment Internal Dimensions (LxHxW): 54.98 x 133.5 x 137.7 mm