Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Racks for 2” Boxes- Rack Only- 2 Boxes Deep x 2 Boxes High
SDP
Catalog No. 503027967
Encompass_Preferred
For Standard 2″ Boxes Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 11 1/2 x 4 1/2 x 5 1/2 inches (292 x 114 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)
Catalog No. 50-302-7967 Supplier Alkali Scientific Supplier No. UFR222
For Standard 2″ Boxes Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-hand doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 11 1/2 x 4 1/2 x 5 1/2 inches (292 x 114 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)