Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Freezer Racks for 3.75″ Cardboard Boxes 3x4 Configuration, 12 Box Capacity
SDP
Catalog No. 503028027
Encompass_Preferred
For Standard 3.75″ Boxes These Upright Freezer Drawer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Constructed of corrosion resistant stainless steel Racks fit standard 3.75" standard boxes Fit all major ultra low (-80°) freezers on the market Dimensions L x H x W: 16 5/8 x 16 11/16 x 5 1/2 inch (286 x 424 x 140mm)Compartment Dimensions L x H x W: 5 3/8 x 3 15/16 x 5 1/4 inch (137 x 100 x 133mm)
Catalog No. 50-302-8027 Supplier Alkali Scientific Supplier No. UFDR34375
Add to Cart
Add to Cart
For Standard 3.75″ Boxes These Upright Freezer Drawer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for an easy removal from the freezer. Constructed of corrosion resistant stainless steel Racks fit standard 3.75" standard boxes Fit all major ultra low (-80°) freezers on the market Dimensions L x H x W: 16 5/8 x 16 11/16 x 5 1/2 inch (286 x 424 x 140mm)Compartment Dimensions L x H x W: 5 3/8 x 3 15/16 x 5 1/4 inch (137 x 100 x 133mm)