Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Rack for 3" Boxes, 5 x 7 Configuration, 35 Box Capacity
SDP
Catalog No. 503028031
Encompass_Preferred
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack 5 Long x 7 Tall= Holds 35 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 27 5/16 x 15 3/8 x 5 1/2 inches (693 x 391 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)
Catalog No. 50-302-8031 Supplier Alkali Scientific Supplier No. UFR572
Constructed of corrosion resistant stainless steel Racks fit standard 2" cardboard, plastic, or metal boxes Security lock devices available for each rack 5 Long x 7 Tall= Holds 35 Boxes  Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 27 5/16 x 15 3/8 x 5 1/2 inches (693 x 391 x 140mm)Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm)