Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Racks for 3.75″ Cardboard Boxes 5x4 Configuration, 20 Box Capacity
SDP
Catalog No. 503028052
Encompass_Preferred
For High Standard 3.75″ Boxes These Upright Freezer Drawer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from the freezer. Constructed of corrosion resistant stainless steel Racks fit standard 3.75" standard boxes Fit all major ultra low (-80°) freezers on the market Dimensions L x H x W: 26 7/8  x 16 11/16 x 5 1/2 inch (683 x 424 x 140mm)Compartment Dimensions L x H x W: 5 3/8 x 3 15/16 x 5 1/4 inch (137 x 100 x 133mm)
Catalog No. 50-302-8052 Supplier Alkali Scientific Supplier No. UFDR54375
For High Standard 3.75″ Boxes These Upright Freezer Drawer Racks for 3.75" standard boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from the freezer. Constructed of corrosion resistant stainless steel Racks fit standard 3.75" standard boxes Fit all major ultra low (-80°) freezers on the market Dimensions L x H x W: 26 7/8  x 16 11/16 x 5 1/2 inch (683 x 424 x 140mm)Compartment Dimensions L x H x W: 5 3/8 x 3 15/16 x 5 1/4 inch (137 x 100 x 133mm)