Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Freezer Drawer Racks for 3″ Cardboard Boxes 2x2 Configuration, 4 Box Capacity
SDP
Catalog No. 503028115
Encompass_Preferred
For Standard 3″ Boxes Our Stainless Steel Freezer Racks with Drawers in a 2 x 2 configuration provides an enhanced storage solution designed for efficiency and ease of use in ultra-low temperature environments. With a capacity to hold six standard 3" boxes, these racks are crafted from high-quality, corrosion-resistant stainless steel, ensuring durability and longevity under the rigorous conditions of ultra-low (-80°C) freezers. Designed for ultra-low upright freezers, these racks feature a drawer system that greatly facilitates the organization, access, and retrieval of samples. Dimensions (DxWxH): 11 3/8 x 5 1/2 x 6 7/16 inches (289 x 140 x 164mm)Compartment Dimensions: 5 3/8 x 5 1/4 x 3 inches (133 x 137.4 x 75.9mm)
Catalog No. 50-302-8115 Supplier Alkali Scientific Supplier No. UFDR223
For Standard 3″ Boxes Our Stainless Steel Freezer Racks with Drawers in a 2 x 2 configuration provides an enhanced storage solution designed for efficiency and ease of use in ultra-low temperature environments. With a capacity to hold six standard 3" boxes, these racks are crafted from high-quality, corrosion-resistant stainless steel, ensuring durability and longevity under the rigorous conditions of ultra-low (-80°C) freezers. Designed for ultra-low upright freezers, these racks feature a drawer system that greatly facilitates the organization, access, and retrieval of samples. Dimensions (DxWxH): 11 3/8 x 5 1/2 x 6 7/16 inches (289 x 140 x 164mm)Compartment Dimensions: 5 3/8 x 5 1/4 x 3 inches (133 x 137.4 x 75.9mm)