Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Freezer Drawer Racks for 2″ Cardboard Boxes 2x6 Configuration, 12 Box Capacity
SDP
Catalog No. 503027818
Encompass_Preferred
For Standard 2″ Boxes Our Stainless Steel Freezer Racks with Drawers in a 2 x 6 configuration provides an enhanced storage solution designed for efficiency and ease of use in ultra-low temperature environments. With a capacity to hold 12 standard 2" boxes, these racks are crafted from high-quality, corrosion-resistant stainless steel, ensuring durability and longevity under the rigorous conditions of ultra-low (-80°C) freezers. Designed for ultra-low upright freezers, these racks feature a drawer system that greatly facilitates the organization, access, and retrieval of samples. Dimensions (DxHxW): 11 3/8  x 14 1/8 x  5 1/2  inches (289  x 359 x 140mm)Compartment Dimensions d x h x w: 5 3/8 x 2 1/16 x 5 1/4  inches (137.4 x 51.6 x 133 mm)
Catalog No. 50-302-7818 Supplier Alkali Scientific Supplier No. UFDR262
For Standard 2″ Boxes Our Stainless Steel Freezer Racks with Drawers in a 2 x 6 configuration provides an enhanced storage solution designed for efficiency and ease of use in ultra-low temperature environments. With a capacity to hold 12 standard 2" boxes, these racks are crafted from high-quality, corrosion-resistant stainless steel, ensuring durability and longevity under the rigorous conditions of ultra-low (-80°C) freezers. Designed for ultra-low upright freezers, these racks feature a drawer system that greatly facilitates the organization, access, and retrieval of samples. Dimensions (DxHxW): 11 3/8  x 14 1/8 x  5 1/2  inches (289  x 359 x 140mm)Compartment Dimensions d x h x w: 5 3/8 x 2 1/16 x 5 1/4  inches (137.4 x 51.6 x 133 mm)