Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless-Steel Freezer Rack for 50-Cell Hinged Top Plastic Boxes, 6 x 3 Configuration, 18 Box Capacity
SDP
Catalog No. 503029045
Encompass_Preferred
For 50-Cell Hinged Top Plastic Boxes The Alkali Scientific™ Upright Stainless Steel Freezer Rack is an essential storage solution engineered to optimize the organization and accessibility of samples stored in 50-cell hinged top plastic boxes within upright freezers. This drawer rack system is designed to enhance the efficiency and capacity of sample storage, making it an invaluable tool for laboratories seeking to maintain high standards of sample integrity and organization Dimensions L x H x W: 21 7/16 x 7 5/8  x 5 7/8 Inch (545 x 193 x 150mm)Compartment Size l x h x w: 3 1/2 x 2 5/16  x 5 9/16 Inch (88.5 x 58 x 142mm)
Catalog No. 50-302-9045 Supplier Alkali Scientific Supplier No. UFDH5063
For 50-Cell Hinged Top Plastic Boxes The Alkali Scientific™ Upright Stainless Steel Freezer Rack is an essential storage solution engineered to optimize the organization and accessibility of samples stored in 50-cell hinged top plastic boxes within upright freezers. This drawer rack system is designed to enhance the efficiency and capacity of sample storage, making it an invaluable tool for laboratories seeking to maintain high standards of sample integrity and organization Dimensions L x H x W: 21 7/16 x 7 5/8  x 5 7/8 Inch (545 x 193 x 150mm)Compartment Size l x h x w: 3 1/2 x 2 5/16  x 5 9/16 Inch (88.5 x 58 x 142mm)