Learn More
Alkali Scientific Stainless Steel Freezer Racks with Drawers for 2" Boxes for PHCbi & NuAire Lab , 4 x 6 Configuration, 24 Box Capacity (Side Access)
SDP
Catalog No. 502388777
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-hand doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer and then sliding the boxes out from either side of the rack. Each drawer of this freezer rack features a slot on the front to insert identification cards for labeling. Dimensions (DxWxH): 22 5/16 x 5 1/2 x 12 5/8 inches (566 x 140 x 321mm). Compartment Dimensions: 5 3/8 x 5 ¼ x 2 1/16 (137.4 x 52 x 133mm).
Catalog No. 50-238-8777 Supplier Alkali Scientific Supplier No. UFDR462S
Racks feature handles at both ends to accommodate freezers with right- or left-hand doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer and then sliding the boxes out from either side of the rack. Each drawer of this freezer rack features a slot on the front to insert identification cards for labeling. Dimensions (DxWxH): 22 5/16 x 5 1/2 x 12 5/8 inches (566 x 140 x 321mm). Compartment Dimensions: 5 3/8 x 5 ¼ x 2 1/16 (137.4 x 52 x 133mm).