Learn More
Alkali Scientific Alkali Scientific™ Upright Stainless Steel Large Freezer Racks for 3.75″ Cardboard Boxes 4x3 Configuration, 12 Box Capacity
SDP
Catalog No. 503027982
Encompass_Preferred
For Large 3.75″ Boxes These Upright Freezer Drawer Racks for 3.75" large boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from the freezer. Constructed of corrosion resistant stainless steel Racks fit standard 3.75" standard boxes Fit all major ultra low (-80°) freezers on the market Dimensions L x H x W: 24 x 12 1/2 x 6 1/8inch (609 x 318 x 155mm)Compartment Dimensions L x H x W: 5 7/8 x 3 15/16 x 5 13/16 inch (149 x 100 x 148mm)
Catalog No. 50-302-7982 Supplier Alkali Scientific Supplier No. UFDR43L375
For Large 3.75″ Boxes These Upright Freezer Drawer Racks for 3.75" large boxes offer convenient storage of samples in an organized format. Constructed of corrosion resistant stainless steel, these freezer racks include durable handles for easy removal from the freezer. Constructed of corrosion resistant stainless steel Racks fit standard 3.75" standard boxes Fit all major ultra low (-80°) freezers on the market Dimensions L x H x W: 24 x 12 1/2 x 6 1/8inch (609 x 318 x 155mm)Compartment Dimensions L x H x W: 5 7/8 x 3 15/16 x 5 13/16 inch (149 x 100 x 148mm)