Learn More
Yamato Scientific America, Inc. Yamato Upright Style Ultra Low Frezer -40ºc To -85ºc, 22 Cu.Ft., Manual Defrost
Catalog No. NC1869584 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Ultra-low upright style freezers comes standard with microprocessor digital controller, audible and visual alarms, remote alarm contacts, alarm battery back-up, and 1/2" access port.
Catalog No. NC1869584 Supplier Yamato Scientific America, Inc. Supplier No. ULF601U
Add to Cart
Add to Cart
Request a Quote
Ultra-low upright style freezers comes standard with microprocessor digital controller, audible and visual alarms, remote alarm contacts, alarm battery back-up, and 1/2" access port.