Learn More
Alkali Scientific Upright Stainless-Steel Freezer Rack for 2" Boxes, 3 x 6 Configuration, 18 Box Capacity
SDP
Catalog No. 502336054
Encompass_Preferred
Standard upright freezer racks for storage of 2 inch boxes, features a 3 boxes deep by 6 boxes high configuration for a total storage of 18 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxHxW): 16 7/8 x 12 5/8 x 5 1/2 inches (429 x 321 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 1/16 x 5 7/16 inches (136 x 52.6 x 138mm).
Catalog No. 50-233-6054 Supplier Alkali Scientific Supplier No. UFR362
Add to Cart
Add to Cart
Standard upright freezer racks for storage of 2 inch boxes, features a 3 boxes deep by 6 boxes high configuration for a total storage of 18 boxes per rack. These upright freezer racks are built with a durable, corrosion resistant stainless steel. Each rack includes handles for an easy retrieval from a laboratory freezer. This freezer rack does not come standard with storage boxes. Dimensions (DxHxW): 16 7/8 x 12 5/8 x 5 1/2 inches (429 x 321 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 1/16 x 5 7/16 inches (136 x 52.6 x 138mm).