Learn More
Alkali Scientific Upright Stainless-Steel Freezer Rack for 2" Boxes, 4 x 6 Configuration, 24 Box Capacity
SDP
Catalog No. 502336130
Encompass_Preferred
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 22 3/16 x 12 3/8 x 5 1/2 inches (429 x 391 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 1/16 x 5 7/16 inches (136 x 52.6 x 138mm).
Catalog No. 50-233-6130 Supplier Alkali Scientific Supplier No. UFR462
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 22 3/16 x 12 3/8 x 5 1/2 inches (429 x 391 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 1/16 x 5 7/16 inches (136 x 52.6 x 138mm).