Learn More
Alkali Scientific Upright Stainless-Steel Freezer Rack for 2" Boxes, 4 x 5 Configuration, 20 Box Capacity
SDP
Catalog No. 502336212
Encompass_Preferred
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers.Dimensions (DxWxH): 22 3/16 x 5 1/2 x 11 inches (564 x 140 x 280mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 2 3/16 inches (137 x 140 x 55mm)
Catalog No. 50-233-6212 Supplier Alkali Scientific Supplier No. UFR452
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers.Dimensions (DxWxH): 22 3/16 x 5 1/2 x 11 inches (564 x 140 x 280mm)Compartment Dimensions: 5 3/8 x 5 7/16 x 2 3/16 inches (137 x 140 x 55mm)