Learn More
TARGETMOL CHEMICALS INC UPROSERTIB 10MG
Supplier: TARGETMOL CHEMICALS INC T684910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Uprosertib (GSK2141795) (GSK2141795) is a selective ATP-competitive and orally bioavailable Akt inhibitor with IC50 of 180 nM 328 nM and 38 nM for Akt 1 2 and 3 respectively. Phase 2. purity: 99%