Learn More
Cole-Parmer UPRT FRZ DRAWER RACK 8-10ML
Supplier:  Cole-Parmer 0440178
Encompass_Preferred
Upright freezer drawer rack for 8-10ml blood sample tubes, 2 drawer, holds 160 tubes, 22 x 9 x 5-1/2
Catalog No. NC0659514