Learn More
Cole-Parmer UPRT.FRZR.DRAWER RACK 20X2"BXS
Catalog No. NC1443272 Shop All Cole-Parmer Products
Encompass_Preferred
freezer racks for 2" boxes (holds 20)26 3/4 x 9 7/16 x 5 1/2 (F-B x H x W)
Catalog No. NC1443272 Supplier Cole-Parmer Supplier No. 0439824
freezer racks for 2" boxes (holds 20)26 3/4 x 9 7/16 x 5 1/2 (F-B x H x W)