Learn More
Thomas Scientific Upright Freezer Rack for 100-Place Slide Boxes, Capacity 16, 28 7/10 x 6 3/10 x 8 2/5" (L x H x W)
SDP
Catalog No. 501459937
Encompass
Stainless Steel Freezer Racks for 100-Place Slide Boxes Argos Technologies® PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Configured to fill all major ultra low (-80°) freezers on the market. Available in both upright and drawer styles. Drawer style allows for easy access to and storage of samples without having to remove the whole rack from the freezer. Both style racks will accommodate a 100-Place slide boxes with dimensions 8.25 x 7.0 x 1.34” (L x W x H) Security Lock devices available for each rack, sold separately
Catalog No. 50-145-9937 Supplier Thomas Scientific Supplier No. 1170X38
Stainless Steel Freezer Racks for 100-Place Slide Boxes Argos Technologies® PolarSafe™ Metal Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors. Configured to fill all major ultra low (-80°) freezers on the market. Available in both upright and drawer styles. Drawer style allows for easy access to and storage of samples without having to remove the whole rack from the freezer. Both style racks will accommodate a 100-Place slide boxes with dimensions 8.25 x 7.0 x 1.34” (L x W x H) Security Lock devices available for each rack, sold separately