Learn More
Grainger UPS sistema alimentación ininterrumpida
Catalog No. NC2608528
Encompass_Preferred
The product is an APC Back-UPS 650VA UPS battery backup and surge protector. It provides battery backup power and surge protection for electronics and computers. Key features include 650VA/390W power capacity, 8 outlets (4 battery backup + 4 surge only), automatic voltage regulation, audible alarms, USB charging port, and LCD display showing power status. The UPS can provide up to 40 minutes of battery power at half load. It has a compact tower design and includes software for power monitoring and safe system shutdown. The UPS is ideal for protecting PCs, networking equipment, and other electronics from power surges and outages.
Catalog No. NC2608528 Supplier Grainger Supplier No. 36HV95
The product is an APC Back-UPS 650VA UPS battery backup and surge protector. It provides battery backup power and surge protection for electronics and computers. Key features include 650VA/390W power capacity, 8 outlets (4 battery backup + 4 surge only), automatic voltage regulation, audible alarms, USB charging port, and LCD display showing power status. The UPS can provide up to 40 minutes of battery power at half load. It has a compact tower design and includes software for power monitoring and safe system shutdown. The UPS is ideal for protecting PCs, networking equipment, and other electronics from power surges and outages.