Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC UPSEM792 hydrochloride | 2341841-08-3 | 99.9% | 277.75 | 25 MG
SDP

Catalog No. 5000394797
Encompass_Preferred

uPSEM792 hydrochloride is a PSAM4-GlyR agonist.

  • Purity: 99.94%
  • Molecular weight: 277.75
  • CAS number: 2341841-08-3
  • Formula: C14H16ClN3O
  • Appearance: Solid
  • Color: White to light yellow
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed, away from moisture and light (solid)
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)

Catalog No. 50-003-94797 Supplier Medchemexpress LLC Supplier No. 25MGHY161182
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

uPSEM792 hydrochloride is a PSAM4-GlyR agonist.

  • Purity: 99.94%
  • Molecular weight: 277.75
  • CAS number: 2341841-08-3
  • Formula: C14H16ClN3O
  • Appearance: Solid
  • Color: White to light yellow
  • Shipping: Room temperature in continental US
  • Storage: 4°C, sealed, away from moisture and light (solid)
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture and light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.