Learn More
Medchemexpress LLC uPSEM792 hydrochloride | 2341841-08-3 | 99.9% | 277.75 g/mol | C14H16ClN3O | 10 MG
SDP
Catalog No. 5000288602
Encompass_Preferred
uPSEM792 hydrochloride is an ultrapotent pharmacologically selective effector molecule (uPSEM) that acts as a high-affinity agonist for engineered PSAM4-GlyR receptors. It is provided for research use only and is commonly used in neuroscience experiments to selectively activate PSAM4-based receptors.
  • High potency agonist for PSAM4-GlyR with reported Ki ≈ 0.7 nM.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as the hydrochloride salt with high chemical purity.
  • Available in small research-scale quantities for experimental use.
Catalog No. 50-002-88602 Supplier Medchemexpress LLC Supplier No. HY16118210MG
uPSEM792 hydrochloride is an ultrapotent pharmacologically selective effector molecule (uPSEM) that acts as a high-affinity agonist for engineered PSAM4-GlyR receptors. It is provided for research use only and is commonly used in neuroscience experiments to selectively activate PSAM4-based receptors.
  • High potency agonist for PSAM4-GlyR with reported Ki ≈ 0.7 nM.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as the hydrochloride salt with high chemical purity.
  • Available in small research-scale quantities for experimental use.