Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Human IKKα

Catalog No. 14461D Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-461-D 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-461-D Supplier MilliporeSigma™ Supplier No. 14461D
Add to Cart
Add to Cart

Expressed in baculovirus-infected Sf9 insect cell expression system.

  • Also known as CHUK, IKK1, IKBKA, TCF16, NFKBIKA
  • Present in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 mg/mL
Product Type Inhibitor
For Use With (Application) Enzyme kinetics, screening inhibitors,selectivity profiling and Kinase Assay
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.