Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Protein A Agarose

Catalog No. 16125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-125 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-125 Supplier MilliporeSigma™ Supplier No. 16125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for purification of human, mouse rabbit immunoglobins.

  • Beads contain sufficient covalently-linked Protein A
  • Binding capacity 20±2 milligrams human IgG/1mm
  • Used in Immunoprecipitation,Western blotting
TRUSTED_SUSTAINABILITY

Specifications

Formulation Water containing 0.01% Thimerosal
For Use With (Application) Purification of human, mouse and rabbit immunoglobins
Content And Storage 4°C
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.