Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Protein A Agarose/Salmon Sperm DNA
GSA_VA

Catalog No. 16157 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-157 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-157 Supplier MilliporeSigma™ Supplier No. 16157
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blocks non-specific DNA binding sites on agarose beads.

EMD Millipore Upstate Protein A Agarose/Salmon Sperm is covalently bound to agarose beads for use in chromatin immunoprecipitations.

  • Protein A covalently bound to agarose by alkylamine linkage
  • Protein A from Staphylococcus aureus has very high affinity for Fc regions of IgG molecules
  • Immobilization of protein A on beads creates an affinity resin which can be used to purify IgG fractions from crude serum, ascites fluid or cell cuture media
  • Qualified using Acetyl Histone H4 H3 ChIP assay
  • Suitable for use in chromatin immunoprecipitation (ChIPs) assays

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Agarose/Salmon Sperm DNA
Formulation 10mM Tris-HCl, 1mM EDTA, pH 8.0 containing 0.05% Sodium Azide.
For Use With (Application) Chromatin immunoprecipitation (ChIPs) assays
Description 2.5mL packed beads
Content And Storage 4°C
Quantity 2.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.