Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Protein G Agarose, Fast Flow

Catalog No. 16266 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-266 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-266 Supplier MilliporeSigma™ Supplier No. 16266
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for medium low pressure chromatography of IgG from mouse, sheep, rabbit, for immunoprecipitations.

EMD Millipore Upstate Protein G Agarose, Fast Flow is stable in all aqueous buffers.

  • Covalently coupled by cyanogen bromide to cross-linked 4% agarose beads
  • Used in Immunoprecipitation, Immunoaffinity purification, Western blotting
TRUSTED_SUSTAINABILITY

Specifications

Formulation Water containing 20% Ethanol
For Use With (Application) Immunoprecipitation, Immunoaffinity Purification, Western Blotting
Description 20mg human IgG/ml agarose
Content And Storage 4°C
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.