Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Protein G Agarose/Salmon Sperm DNA

Catalog No. 16201 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-201 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-201 Supplier MilliporeSigma™ Supplier No. 16201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blocks non-specific DNA binding sites on agarose beads.

EMD Millipore Upstate Protein G Agarose/Salmon Sperm DNA is covalently bound to agarose beads by cyanogen bromide linkage for use in chromatin immunoprecipitations.

  • Covalently bound to agarose by cyanogen bromide linkage
  • Has very high affinity for the Fc regions of IgG molecules
  • Immobilization of protein G on beads creates affinity resin which is used to purify IgG fractions from crude serum, ascites fluid or cell cuture media
  • 2.5mL packed beads contains 1mg sonicated salmon sperm DNA, 2.5mg BSA approximately 5mg recombinant Protein G
  • Used in chromatin immunoprecipitations (ChIP assays)
  • Qualified using Acetyl Histone H3 ChIP assay
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Chromatin immunoprecipitation (ChIPs) assays
Description 2.5mL packed beads
Content And Storage 4°C
Quantity 2.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.