Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Upstate Streptavidin, Agarose Conjugate

Catalog No. 16126 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-126 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-126 Supplier MilliporeSigma™ Supplier No. 16126
Add to Cart
Add to Cart

Used for purification of biotinylated macromolecules.

  • Isolated from Streptomyces avidinii
  • Covalently bound to agarose
  • Used in Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Formulation PBS containing 0.02% Sodium Azide
Content And Storage 4°C
Quantity 10 mL
For Use With (Application) Suitable for purification of biotinylated macromolecules, Immunoprecipitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.