Learn More
MSC Nilfisk Industrial Cleaner, Electric, HEPA Filter, 13 gal Capacity, Steel Tank
Catalog No. 502838184
Encompass
This single-phase industrial vacuum is designed for diverse cleaning challenges across industries including food, paper, packaging, powder coating, textiles, and woodworking. It offers robust construction and advanced innovations.
  • Modular design for easy modifications and servicing.
  • Electronic keypad with LED indicators to monitor filter performance.
  • Manual filter shaker with polyester main filter for high filtration efficiency.
  • Upstream HEPA filtration ensures 99.995% of particles are retained, down to 0.18 microns.
  • Polyliner adapter kit for easy disposal of collected debris.
  • Includes a 50 mm accessory kit with 20 ft antistatic hose, double-curved wand, 16 in wheeled floor nozzle, 20 in metal crevice nozzle, and container polyliners.
Catalog No. 50-283-8184 Supplier MSC Supplier No. 38686614
Add to Cart
Add to Cart
Request a Quote
This single-phase industrial vacuum is designed for diverse cleaning challenges across industries including food, paper, packaging, powder coating, textiles, and woodworking. It offers robust construction and advanced innovations.
  • Modular design for easy modifications and servicing.
  • Electronic keypad with LED indicators to monitor filter performance.
  • Manual filter shaker with polyester main filter for high filtration efficiency.
  • Upstream HEPA filtration ensures 99.995% of particles are retained, down to 0.18 microns.
  • Polyliner adapter kit for easy disposal of collected debris.
  • Includes a 50 mm accessory kit with 20 ft antistatic hose, double-curved wand, 16 in wheeled floor nozzle, 20 in metal crevice nozzle, and container polyliners.