Learn More
Formulatrix μPULSE® v3 Tube Adapter – 15 ml, THIS ACCESSORY IS ONLY COMPATIBLE WITH µPULSE V3 SYSTEMS (Serial No. 221 and higher).
SDP
Catalog No. NC2751039
Encompass
μPULSE® v3 Tube Adapter – 15 mlTHIS ACCESSORY IS ONLY COMPATIBLE WITH µPULSE V3 SYSTEMS (Serial No. 221 and higher).This Tube Adapter holds a standard 15mL conical tube. This part interfaces the standard 50mL Tube Adapter to accommodate the smaller 15mL tubes.
Catalog No. NC2751039 Supplier Formulatrix Supplier No. 815247
μPULSE® v3 Tube Adapter – 15 mlTHIS ACCESSORY IS ONLY COMPATIBLE WITH µPULSE V3 SYSTEMS (Serial No. 221 and higher).This Tube Adapter holds a standard 15mL conical tube. This part interfaces the standard 50mL Tube Adapter to accommodate the smaller 15mL tubes.