Learn More
Labrepco Inc UPWRIGHT FRZR RCK W/DRWS
SDP
Catalog No. NC2079327
Encompass_Preferred
Value Series Upright Freezer Rack with Drawers for Standard 2″ Boxes- Rack Only (3 Boxes Deep x 3 Boxes High)
Catalog No. NC2079327 Supplier Labrepco Inc Supplier No. CUFT332
Value Series Upright Freezer Rack with Drawers for Standard 2″ Boxes- Rack Only (3 Boxes Deep x 3 Boxes High)