Learn More
Cayman Chemical UR-144 Degradant, Purity: ≥98%, CAS: 1609273-88-2, 10 mg
SDP
Catalog No. 502954604
Encompass_Preferred
A common impurity observed during GC-MS analysis of samples containing UR-144, featuring an opened ring presumed to be produced during heating; gives rise to a prominent fragment ion that is 15 amu greater than the base peak of UR-144
Catalog No. 50-295-4604 Supplier Cayman Chemical Supplier No. 1192810 MG
A common impurity observed during GC-MS analysis of samples containing UR-144, featuring an opened ring presumed to be produced during heating; gives rise to a prominent fragment ion that is 15 amu greater than the base peak of UR-144