Learn More
Cayman Chemical UR144 Degradnt NpentnoIc 10mg
SDP
Catalog No. 502892473
Encompass_Preferred
A potential phase I metabolite of a degradation product observed during GC-MS analysis of samples containing UR-144; intended for forensic and research applications
Catalog No. 50-289-2473 Supplier Cayman Chemical Supplier No. 900145310 MG
A potential phase I metabolite of a degradation product observed during GC-MS analysis of samples containing UR-144; intended for forensic and research applications