Learn More
Cayman Chemical UR-144 N-(5-bromopentyl) analog, Purity: ≥98%, CAS: 1628690-26-5, 10 mg
SDP
Catalog No. 502955792
Encompass_Preferred
Differs from UR-144 by having a bromine atom on the terminal carbon of the alkyl group; intended for research and forensic applications
Catalog No. 50-295-5792 Supplier Cayman Chemical Supplier No. 1200310 MG
Differs from UR-144 by having a bromine atom on the terminal carbon of the alkyl group; intended for research and forensic applications