Learn More
Cayman Chemical UR-144 N-(5-chloropentyl) analog, Purity: ≥98%, CAS: 1445577-42-3, 10 mg
SDP
Catalog No. 502953236
Encompass_Preferred
Differs from UR-144 by having a chlorine atom on the terminal carbon of the alkyl group; intended for research and forensic applications
Catalog No. 50-295-3236 Supplier Cayman Chemical Supplier No. 1195110 MG
Differs from UR-144 by having a chlorine atom on the terminal carbon of the alkyl group; intended for research and forensic applications