Learn More
Cayman Chemical UR144 N 5hydroxypentyl me 10mg
SDP
Catalog No. 502952393
Encompass_Preferred
An expected phase I metabolite of UR-144, based on the metabolism of similar cannabimimetics; should be detectable in either serum or urine; intended for forensic and research applications
Catalog No. 50-295-2393 Supplier Cayman Chemical Supplier No. 1177510 MG
An expected phase I metabolite of UR-144, based on the metabolism of similar cannabimimetics; should be detectable in either serum or urine; intended for forensic and research applications