Learn More
Cayman Chemical UR-144 N-pentanoic acid metabolite, Purity: ≥98%, CAS: 1451369-33-7, 10 mg
SDP
Catalog No. 502952359
Encompass_Preferred
An expected phase I metabolite of UR-144; should be detectable in either serum or urine; intended for forensic and research applications
Catalog No. 50-295-2359 Supplier Cayman Chemical Supplier No. 1177310 MG
An expected phase I metabolite of UR-144; should be detectable in either serum or urine; intended for forensic and research applications