Learn More
Selleck Chemical LLC Uracil 1-b-D-arabinofuranoside S4807-25mg
SDP
Catalog No. 503108146
Encompass_Preferred
Uracil 1- -D-arabinofuranoside (Spongouridine Arauridine 1-beta-D-Arabinofuranosyluracil Ara-U) is a deamination metabolite of Cytarabine
Catalog No. 50-310-8146 Supplier Selleck Chemical LLC Supplier No. S480725MG
Uracil 1- -D-arabinofuranoside (Spongouridine Arauridine 1-beta-D-Arabinofuranosyluracil Ara-U) is a deamination metabolite of Cytarabine