Learn More
Cayman Chemical UracIl-13C 15N2 5mg
SDP
Catalog No. 502947496
Encompass_Preferred
An internal standard for the quantification of uracil by GC- or LC-MS
Catalog No. 50-294-7496 Supplier Cayman Chemical Supplier No. 339975 MG
An internal standard for the quantification of uracil by GC- or LC-MS